Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Propranolol-D7 (ring-D7) Solution, 100 μg/mL in Methanol with 5% 1 M HCl, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001859 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-859 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-859 Supplier MilliporeSigma Supelco Supplier No. P0851ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Propranolol is a sympatholytic, non-selective beta blocker used to treat hypertension, anxiety and panic. The drug is sold under numerous brand names such as Inderal™, Avlocardyl, Deralin, and Dociton. This internal standard is suitable for quantitation of propranolol levels in patient samples using LC/MS or GC/MS for applications in clinical toxicology, diagnostic testing, forensic analysis, pharmaceutical research and isotope dilution methods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 344298-99-3
Linear Formula C16H14D7NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol with 5% 1 M HCl
Synonym Propranolol-d7 (ring-d7)
Formula Weight 266.39
Packaging Ampule
Quantity 1 mL
Flash Point 11°C (51.8°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.