Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Protriptyline-d3 Hydrochloride, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001862 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-862 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-862 Supplier MilliporeSigma Supelco Supplier No. P0881ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A stable labeled internal standard for protriptyline testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, or urine drug testing. Protriptyline is a second generation tricyclic antidepressant (TCA) marketed under the trade name Vivactil™, for treatment of major depression and ADHD.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C19D3H18N · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Synonym Protriptyline-D3 hydrochloride solution
Formula Weight 302.86
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.