Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Pyridoxal 5'-phosphate Solution, 500 μg/mL (80:20 Water:Acetonitrile), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002179 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-179 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-179 Supplier MilliporeSigma Supelco Supplier No. V0691ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Pyridoxal 5′-phosphate, also known as PLP or P5P, is the active form of vitamin B6 and acts as coenzyme in a variety of enzymatic reactions. This certified solution standard is suitable for use as starting materials in the preparation of calibrators, controls, or linearity standards in a variety of testing applications from diagnostic vitamin B testing to quality control testing of dietary supplements, fortified foods or pharmaceutical preparations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 54-47-7
Linear Formula C8H10NO6P
MDL Number MFCD00006333
UN Number UN 1648 3 / PGII
Concentration 500 μg/mL (80:20 water:acetonitrile)
Formula Weight 247.14
Packaging Ampule
Quantity 1 mL
Flash Point 17°C (62.6°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.