Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Quetiapine fumarate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001914 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-914 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-914 Supplier MilliporeSigma Supelco Supplier No. Q0011ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Quetiapine, marketed as Seroquel™, is an atypical antipsychotic drug for the treatment of depression, bipolar disorder, and schizophrenia. This Snap-N-Spike™ Reference Solution is applicable to numerous GCMS and LCMS applications including forensic toxicology analysis, calibrator preparation and urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 111974-72-2
Linear Formula C21H25N3O2S · 0.5 C4H4O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Formula Weight 441.54
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.