Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (R,R)-(-)-Pseudoephedrine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001835 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-835 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-835 Supplier MilliporeSigma Supelco Supplier No. P0361ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Pseudoephedrine, a phenethylamine analog, is used as a decongestant in over-the-counter cough and cold preparations such as Sudafed, Claritin-D, and MucinexD. Pseudoephedrine is a reagent in illegal methamphetamine production and is used by athletes as a performance enhancing drug. This Snap-N-Spike™ standard is suitable for LC/MS or GC/MS pseudoephedrine testing applications in forensic analysis, urine drug testing, clinical toxicology and sports testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 321-97-1
Linear Formula C10H15NO
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Synonym (1R,2R)-(-)-Pseudoephedrine
Formula Weight 165.23
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.