Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ RDX Solution, 1000 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco ERR001S1.2ML

Catalog No. 01-001-339


Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 121-82-4
MFCD00458725
1000 μg/mL in acetonitrile
222.12
1.2 mL
C3H6N6O6
UN 1648 3 / PGII
Hexahydro-1,3,5-trinitro-1,3,5-triazine, Hexahydro-1,3,5-trinitro-1,3,5-triazine solution
Ampule
2°C (35.6°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H332-H319

H312

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation