Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Remifentanil-13C6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001934 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-934 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-934 Supplier MilliporeSigma Supelco Supplier No. R0340.5ML

missing translation for 'validMainframeMsg'

Certified reference material

Remifentail (Ultivasup) is a potent, short-acting, synthetic opioid analgesic drug. It is used in combination with other opioids and analgesics to induce or supplement general anesthesia. An internal standard suitable for remifentanyl testing of urine, blood, or hair samples or isotope dilution methods by GC/MS or LC/MS in applications such as pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C1413C6H28N2O5 · HCl
MDL Number MFCD09952283
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 418.86
Packaging Ampule
Quantity 0.5 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.