Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (-)-Riboflavin, 100 μg/mL (1% ammonium acetate in 50:50 Methanol:Water), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002177 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-177 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-177 Supplier MilliporeSigma Supelco Supplier No. V0671ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Riboflavin is an essential B vitamin for maintaining human health with key roles in energy metabolism as well as a critical component of enzyme cofactors and flavoproteins.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 83-88-5
Melting Point 290°C (dec.)
Linear Formula C17H20N4O6
UN Number UN 1992 6.1(3) / PGIII
Concentration 100 μg/mL (1% ammonium acetate in 50:50 methanol:water)
Synonym Lactoflavin, Vitamin B2, Vitamin G
Formula Weight 376.36
Packaging Ampule
Quantity 1 mL
Flash Point 24°C (75.2°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.