Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Risperidone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco R0061ML

Catalog No. 01-001-919


Add to Cart

Description

Description

Risperidone, marketed as Risperdal™, Ridal, or Riscalin, is an atypical antipsychotic used to treat schizophrenia, bipolar disorder, and irritability in autistic children. This new Certified Snap-N-Spike™ Solution is suitable for many LC/MS and GC/MS applications including forensic analysis, clinical toxicology, or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 106266-06-2
UN 1230 - class 3 - PG 2 - Methanol, solution
410.48
1 mL
C23H27FN4O2
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation