Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (S)-N-Nitrosoanatabine Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001745 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-745 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-745 Supplier MilliporeSigma Supelco Supplier No. N0781ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

N-Nitrosoanatabine or NAT is a potent carcinogen and biomarker of exposure to cigarette smoke. It's found in a variety of tobacco products such as cigarettes, electronic cigarettes, nicotine gum and oral snuff. This certified Snap-N-Spike™ solution is suitable for use in LC-MS/MS or GC/MS methods for nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 71267-22-6
Linear Formula C10H11N3O
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Synonym NAT
Formula Weight 189.21
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.