Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Salvinorin A Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Salvinorin A, listed on the DEA's Drugs and Chemicals of Concern website, is the main psychotropic agent of the hallucinogenic drug Salvia also known as Sally-D, Magic Mint, and Seer's Sage. Salvinorin A is considered the most potent naturally occurring hallucinogen.
Specifications
Specifications
| CAS | 75-05-8, 83729-01-5 |
| Linear Formula | C23H28O8 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 432.46 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?