Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Secobarbital-D5 Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001961 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-961 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-961 Supplier MilliporeSigma Supelco Supplier No. S0481ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for secobarbital testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, pain prescription monitoring, or urine drug testing. Secobarbital, sold under the trade name Seconal™, is a barbiturate used in the treatment of epilepsy, the short term treatment of insomnia and as a preoperative anesthetic.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 145243-97-6
Linear Formula C12D5H13N2O3
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 243.31
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.