Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Sibutramine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco S0111ML

Catalog No. 01-001-939


Add to Cart

Description

Description

Sibutramine, a SNRI and anorectant, was marketed as a treatment for obesity until 2010 when it was removed from the market over concerns of potential adverse cardiovascular side effects. Sold under trade names Reductil, Meridia™, and Subitrex, this sibutramine Snap-N-Spike™ reference solution is applicable for use in LC/MS or GC/MS applications for clinical toxicology and forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 84485-00-7
MFCD04039806
1.0 mg/mL in methanol (as free base)
316.31
1 mL
C17H26ClN · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
Sibutramine hydrochloride monohydrate, 1-(4-Chlorophenyl)-N,N-dimethyl-alpha-(2-methylpropyl)cyclobutanemethanamine hydrochloride monohydrate, Meridia, Reductil
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation