Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Sirolimus Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001941 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-941 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-941 Supplier MilliporeSigma Supelco Supplier No. S0151ML
Add to Cart
Add to Cart

Certified reference material

A Certified Spiking Solution™ of an immunosuppressant and narrow therapeutic index drug used to prevent rejection in organ transplantation. This certified calibration material is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug's narrow therapeutic range. The Snap-N-Spike™ format allows laboratories to eliminate weighing operations of hazardous substances such as sirolimus, significantly reducing occupational exposure hazards related to handling of acutely toxic powders, and provides the ability to spike into their matrix of choice just before use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 53123-88-9
Linear Formula C51H79NO13
MDL Number MFCD00867594
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Synonym Rapamycin
Formula Weight 914.17
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.