Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Spice Cannabinoid Mix Solution, 100 μg/mL each component in acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01258406 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-406 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-406 Supplier MilliporeSigma Supelco Supplier No. S0381ML
Add to Cart
Add to Cart

A multi-component mix containing Spice cannabinoids JWH-250; JWH-200; HU-211; (±)-CP 47,497; and (±)-CP 47,497 C8 homologue.

On March 1, 2011, the US Drug Enforcement Agency (DEA) used its emergency power to classify five synthetic cannabinoids in Schedule I, the most restrictive category under the Controlled Substances Act. The five synthetic cannabinoids under this one year ban include JWH-018, JWH-073, JWH-200, CP-47,497 and its C8 homologue.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampoule
Product Line CERILLIANT
Physical Form Liquid
Concentration 100 μg/mL each component in Acetonitrile
Industry Type Forensics, Toxicology
Quantity 1 mL
Grade Certified Reference Material

CERILLIANT™ is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.