Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tapentadol-beta-D-glucuronide Solution, 100 μg/mL in Acetonitrile:Water, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0601ML

Catalog No. 01-002-062


 Restricted Item
Add to Cart

Description

Description

The first Certified Spiking Solution™ of the primary urinary metabolite of tapentadol. Tapentadol is the first new drug of the centrally-acting analgesic class approved in the US for pain management in more than 25 years. This Certified Spiking Solution™ is suitable for many LC/MS or GC/MS applications, including pain prescription monitoring, forensic testing, clinical toxicology, or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8 , 1300037-86-8
MFCD21363883
100 μg/mL in acetonitrile:water
Ampule
13°C (55.4°F) - closed cup
C20H31NO7
UN 1648 3 / PGII
397.46
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation