Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tapentadol Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0581ML

Catalog No. 01-002-060


Add to Cart

Description

Description

Tapentadol represents the first new drug of the centrally-acting analgesic class approved in the US in more than 25 years. This Snap-N-Spike™ Reference Solution is suitable for many LC/MS or GC/MS applications, including forensic analysis, pharmaceutical analysis, pain prescription monitoring, and urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 175591-09-0
MFCD00944992
1.0 mg/mL in methanol (as free base)
257.80
1 mL
C14H23NO · HCl
UN 1230 - class 3 - PG 2 - Methanol, solution
Tapentadol hydrochloride solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation