Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Temazepam-d5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T9021ML

Catalog No. 01-002-114


 Restricted Item
Add to Cart

Description

Description

Temazepam-D5 is a stable-labeled internal standard suitable for quantitation of temazepam levels or isotope dilution methods by GC/MS or LC/MS for applications such as clinical toxicology, urine drug testing, and pain prescription monitoring. Temazepam is a benzodiazepine drug approved for the short term treatment of insomnia and sold under the trade names Restoril™ and Normison.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 136765-51-0
MFCD00673311
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C16D5H8ClN2O2
UN 1230 - class 3 - PG 2 - Methanol, solution
305.77
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation