Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Temazepam glucuronide lithium salt Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002059 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-059 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-059 Supplier MilliporeSigma Supelco Supplier No. T0501ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

The major urinary and blood metabolite of temazepam, an intermediate-acting 3-hydroxy benzodiazepine prescribed for the short-term treatment of sleeplessness in patients with difficulties remaining asleep. This new Certified Reference Standard is suitable for a variety of LCMS and GCMS applications, including forensic and forensic toxicology analysis, urine drug testing, and hydrolysis controls.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C22H20ClLiN2O8
MDL Number MFCD29918103
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free carboxylate)
Formula Weight 482.80
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.