Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tetrahydrocannabivarin (THCV) Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002083 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-083 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-083 Supplier MilliporeSigma Supelco Supplier No. T0941ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Tetrahydrocannabivarin (THCV) is a cannabinoid found in Cannabis that reportedly has efficacy for the treatment of diabetes and controlling weight gain. This certified Snap-N-Spike™ solution is suitable for THCV testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling and pharmaceutical research. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C19H26O2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 286.41
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.