Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tetrazepam Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002094 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-094 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-094 Supplier MilliporeSigma Supelco Supplier No. T1241ML
 Restricted Item
Add to Cart
Add to Cart

Certified reference material

Tetrazepam, marketed under brand names such as Clinoxan, Epsipam, Myolastan, Musaril, Relaxam and Spasmorelax, is a benzodiazepine derivative with anticonvulsant, anxiolytic, muscle relaxant and slightly hypnotic properties. Tetrazepam is used to treat muscle spasm, anxiety disorders, depression, premenstrual syndrome or agoraphobia. This Certified Spiking Solution™ is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 10379-14-3
Linear Formula C16H17ClN2O
Concentration 1 mg/mL in acetonitrile
Formula Weight 288.77
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.