Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Thebaine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T1161ML

Catalog No. 01-002-087


Add to Cart

Description

Description

Thebaine is a natural constituent of opium that is chemically similar to both morphine and codeine, but exerts stimulatory rather than depressant effects. Analysis of thebaine maybe performed to rule-out or confirm a false morphine positive. This Certified Snap-N-Spike™ Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 115-37-7
MFCD00027409
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F)
C19H21NO3
UN 1230 - class 3 - PG 2 - Methanol, solution
311.37
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation