Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Thioridazine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T9051ML

Catalog No. 01-002-116


Add to Cart

Description

Description

An analytical reference standard applicable for use as starting material in calibrators or controls for thioridazine testing methods by LC-MS/MS or GC-MS. Thioridazine is a treatment for schizophrenia sold under trade names Mellaril, Novoridazine, and Thioril.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 50-52-2
MFCD00242875
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C21H26N2S2
UN 1230 - class 3 - PG 2 - Methanol, solution
370.57
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370-H412

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation