Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-threo-Ritalinic acid-d10 Hydrochloride, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco R0141ML

Catalog No. 01-001-922


 Restricted Item
Add to Cart

Description

Description

Ritalinic acid is the major urinary metabolite of methylphenidate, more commonly known by the brand name Ritalin™ which is approved for treatment of ADHD (Attention-Deficit Hyperactivity Disorder) and narcolepsy. This new stable-labeled internal standard is suitable for a wide variety of GC/MS and LC/MS applications from clinical toxicology and urine drug testing to forensic analysis and isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
(+/-)-threo-Ritalinic acid-d10 HCl solution
Ampule
9.7°C (49.5°F) - closed cup
C13H7NO2D10
100 μg/mL in methanol (as free base)
229.34
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation