Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Tilidine-D6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002066 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-066 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-066 Supplier MilliporeSigma Supelco Supplier No. T0691ML
Add to Cart
Add to Cart

Certified reference material

This stable-labeled internal standard is suitable for quantification of tilidine levels in GC/MS or LC/MS testing methods for clinical toxicology, forensic analysis, urine drug testing, or isotope dilution applications. Tilidine is a synthetic opioid analgesic used mainly in Germany, Switzerland and Belgium for treatment of moderate to severe pain, both acute and chronic. While tilidine is considered a moderately-potent opioid, its active metabolite nortilidine possesses analgesic effects roughly equivalent to that of morphine.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C17D6H17NO2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 315.87
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.