Learn More
CERILLIANT™ Topiramate-D12 Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco T1501ML
Catalog No. 01-002-107
Add to Cart
Description
Description
Suitable uses for this certified solution standard include as an internal standard for quantitation of topiramate levels in biological matrices such as urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Topiramate, sold under trade name Topamax™, is an antiepileptic drug used for the treatment of seizures. Topiramate in combination with the stimulant phentermine constitute the active compounds in the new weight lose drug Qsymia™.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
67-56-1 , 1279037-95-4
MFCD09752150
351.44
1 mL
C12D12H9NO8S
1 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Product Title
Select an issue
CERILLIANT is a registered trademark of Cerilliant Corporation