Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Topiramate-D12 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco T0411ML

Catalog No. 01-002-053


Add to Cart

Description

Description

Suitable uses for this certified solution standard include as an internal standard for quantitation of topiramate levels in biological matrices such as urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Topiramate, sold under trade name Topamax™, is an antiepileptic drug used for the treatment of seizures. Topiramate in combination with the stimulant phentermine constitute the active compounds in the new weight lose drug Qsymia™.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1279037-95-4
MFCD09752150
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C12D12H9NO8S
UN 1230 - class 3 - PG 2 - Methanol, solution
351.44
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation