Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ trans-3'-Hydroxycotinine-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001457 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-457 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-457 Supplier MilliporeSigma Supelco Supplier No. H1081ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Trans-3'-Hydroxycotinine is a major nicotine metabolite excreted in the urine of smokers and users of other tobacco or nicotine products including cigars, chewing tobacco and snuff. This certified Snap-N-Spike™ solution is suitable for use as an internal standard for quantification of trans-3'-Hydroxycotinine levels in urine or other matrices for nicotine testing by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 159956-78-2
Linear Formula C10H9D3N2O2
UN Number UN 1230 - class 3 - PG 2 - Methanol
Concentration 100 μg/mL in methanol
Formula Weight 195.23
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.