Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ trans-3'-Hydroxycotinine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001451 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-451 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-451 Supplier MilliporeSigma Supelco Supplier No. H1011ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Trans-3'-Hydroxycotinine is a major nicotine metabolite excreted in the urine of smokers and users of other tobacco or nicotine products including cigars, chewing tobacco and snuff. This Certified Spiking Solution™ is suitable for use in nicotine testing methods or clinical and diagnostic testing of nicotine biomarkers by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 34834-67-8
Linear Formula C10H12N2O2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 192.21
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.