Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Trinitroglycerin Solution, 1 % (w/w) in propylene glycol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002042 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-042 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-042 Supplier MilliporeSigma Supelco Supplier No. T0211ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Trinitroglycerin, also commonly known as nitroglycerin, is a pharmaceutical vasodilator used to treat heart conditions such as angina and chronic heart failure. This explosive compound is also an ingredient in dynamite and gunpowder. This Certified Spiking Solution™ is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology.
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-55-6, 55-63-0
Linear Formula C3H5N3O9
MDL Number MFCD00171649
UN Number UN 1204 - class 3 - PG 2 - Nitroglycerin solution in alcohol
Concentration 1% (w/w) in propylene glycol
Formula Weight 227.09
Packaging Ampule
Quantity 1 mL
Flash Point 11°C (51.8°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.