Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ UR-144 5-Hydroxypentyl metabolite-D5 (indole-D5) Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001981 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-981 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-981 Supplier MilliporeSigma Supelco Supplier No. S0791ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

An internal standard suitable for quantitation of UR-144 5-Hydroxypentyl metabolite levels using LC-MS/MS or GC/MS for synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. UR-144 5-Hydroxypentyl metabolite is a primary urinary metabolite of UR-144, a third generation synthetic cannabinoid that has seen wide popularity in illicit drug markets since late 2012. This biomarker is also referred to as the terminal omega hydroxyl metabolite of UR-144.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Linear Formula C21H24D5NO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 332.49
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.