Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ UR-144 5-Hydroxypentyl metabolite Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco S0771ML

Catalog No. 01-001-979


 Restricted Item
Add to Cart

Description

Description

A certified solution standard suitable for use as starting material in calibrators and controls for GC/MS or LC/MS synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. This monohydroxy analog is a primary urinary metabolite of UR-144, a synthetic cannabinoid found in blends of the herbal mixture K2 or Spice. This biomarker is also referred to as the terminal omega hydroxyl metabolite of UR-144.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 895155-95-0
UN 1230 - class 3 - PG 2 - Methanol
327.46
1 mL
C21H29NO2
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation