Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ UR-144 5-Pentanoic acid metabolite Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001980 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-980 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-980 Supplier MilliporeSigma Supelco Supplier No. S0781ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A primary urinary metabolite of UR-144, a synthetic cannabinoid found in many blends of the herbal mixture Spice also known as K2, Genie, or Demon. For controlled synthetic cannabinoids Cerilliant offers DEA-exempt certified solutions, allowing laboratories to order these standards without DEA paperwork.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1451369-33-7
Linear Formula C21H27NO3
UN Number UN 1230 - class 3 - PG 2 - Methanol
Concentration 100 μg/mL in methanol
Formula Weight 341.44
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.