Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Valproic acid Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002136 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-136 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-136 Supplier MilliporeSigma Supelco Supplier No. V0061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A Certified Spiking Solution™ suitable for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, urine drug testing, prescription monitoring or pharmaceutical research. Valproic acid, sold under trade names including Depakote™, Valparin, or Stavzor™, is an antiepileptic drug and mood stabilizer. The drug is prescribed for treatment of epilepsy, bipolar disorder, migraine headaches, and depression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 99-66-1
Linear Formula C8H16O2
MDL Number MFCD00002672
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 144.21
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.