Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Venlafaxine-D6 Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco V0371ML

Catalog No. 01-002-160


Add to Cart

Description

Description

Also known by the brand name Effexor™, venlafaxine is an SNRI antidepressant approved for the treatment of major depressive and general anxiety disorders. This stable-labeled internal standard is suitable for many LC/MS and GC/MS applications from forensic or clinical toxicology to urine drug testing and isotope dilution methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 1062606-12-5
UN 1230 - class 3 - PG 2 - Methanol, solution
319.90
1 mL
C17D6H21NO2 · HCl
1.0 mg/mL in methanol (as free base)
Ampule
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation