Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Venlafaxine-D6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002138 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-138 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-138 Supplier MilliporeSigma Supelco Supplier No. V0091ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

A new internal standard suitable for many LC/MS and GC/MS applications from forensic or clinical toxicology analysis to urine drug testing and isotope dilution methods. Also known by the brand name Effexor™, venlafaxine is an SNRI antidepressant approved for the treatment of major depressive and general anxiety disorders.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1062606-12-5
Linear Formula C17D6H21NO2 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 319.90
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.