Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Vitamin K2 (MK-4)-13C6 Solution, 10 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
This stable-labeled internal standard is suitable for quantification of vitamin K2 (MK-4) in LC-MS/MS clinical testing applications such as assessment of vitamin K2 defficiency and research of VKD proteins. Vitamin K2 (MK-4) or menaquinone-4 is an essential cofactor for enzymes that activate blood-clotting factors as well as vitamin K-dependent (VKD) proteins involved in bone metabolism, arterial calcification, growth control, and signal transduction.
Specifications
Specifications
| CAS | 75-05-8 |
| Linear Formula | C2513C6H40O2 |
| MDL Number | MFCD00079646 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 10 μg/mL in acetonitrile |
| Formula Weight | 30327.27 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?