Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ XLR-11 4-Hydroxypentyl metabolite Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco S0801ML

Catalog No. 01-001-982


Add to Cart

Description

Description

A primary urinary metabolite of XLR-11, a popular third generation synthetic cannabinoid that purportedly can cause serious adverse health effects including kidney damage and stroke. XLR-11 is similar in structure to UR-144 and A-796,260, two compounds originally developed as pharmaceutical drug candidates due to their high affinity for the cannabinoid receptor CB2. Our certified synthetic cannabinoid solution standards are DEA-exempt, allowing laboratories to order these standards without DEA paperwork.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
345.45
1 mL
C21H28FNO2
100 μg/mL in methanol
Ampule
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation