Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ XLR-11 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001973 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-973 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-973 Supplier MilliporeSigma Supelco Supplier No. S0611ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

XLR-11 is a synthetic cannabinoid of the tetramethylcyclopropyl series found in newer Spice blends. Along with UR-144, cyclopropyl Spice analogs have seen wide popularity in the illicit markets since late 2012. XLR-11 is similar in structure to UR-144 and A-796,260, two compounds originally developed as pharmaceutical drug candidates due to their high affinity for the cannabinoid receptor CB2.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 1364933-54-9
Linear Formula C21H28FNO
MDL Number MFCD23701752
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 329.45
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F)

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.