Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Zaleplon Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco Z0041ML

Catalog No. 01-002-188


Add to Cart

Description

Description

Zaleplon is a nonbenzodiazepine 'Z-drug' prescribed under the trade names Sonata™ or Starnoc for the treatment of insomnia. Zaleplon is often abused illicitly for its mind-altering effects. This Certified Spiking Solution™ is applicable to use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 151319-34-5
UN 1230 - class 3 - PG 2 - Methanol, solution
305.33
1 mL
C17H15N5O
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation