Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Zolazepam Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002200 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-200 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-200 Supplier MilliporeSigma Supelco Supplier No. Z0201ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

Zolazepam, also known as Flupyrazapon, is a pyrazolodiazepinone derivative structurally related to benzodiazepine drugs. Zolazepam, usually used as anesthetic in veterinary medicine, recently appeared on the illicit drug market. A Certified Snap-N-Spike™ Solution suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, and urine drug testing.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 33754-49-3
Linear Formula C15H15FN4O · HCl
MDL Number MFCD01940774
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1 mg/mL in methanol (as free base)
Formula Weight 322.77
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.