Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Zolpidem-D7 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco Z0081ML

Catalog No. 01-002-192


Add to Cart

Description

Description

Zolpidem, marketed as Ambien™ or Stilnox™, is a nonbenzodiazepine used for the short-term treatment of insomnia. This stable-labeled internal standard is uniquely suited for GC/MS applications, because unlike the dimethyl-D6 analog, the deuterium label is retained in the major product ion fragment. As with dimethyl-D6 internal standard, the D7-labeled analog is suitable for LC/MS methods.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
Zolpidem-D7 (Suitable for use with GC/MS or LC/MS) solution
Ampule
9.7°C (49.5°F) - closed cup
C19H14N3OD7
100 μg/mL in methanol
314.43
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation