Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Zolpidem Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco Z0171ML

Catalog No. 01-002-197


Add to Cart

Description

Description

Zolpidem is a non-benzodiazepine hypnotic sold under the trade names Ambien™, Stilnox™ and Edluar™ for the treatment of insomnia. This Certified Snap-N-Spike™ Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing. Zolpidem has a wide history of abuse, including recreational use and as a date-rape drug.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1 , 82626-48-0
MFCD00153885
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C19H21N3O
UN 1230 - class 3 - PG 2 - Methanol, solution
307.39
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

CERILLIANT is a registered trademark of Cerilliant Corporation