Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Zopiclone-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002201 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-002-201 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-002-201 Supplier MilliporeSigma Supelco Supplier No. Z9021ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material

This internal standard is suitable for many LC/MS and GC/MS applications including forensic analysis, clinical toxicology, urine drug testing, or isotope dilution methods. Zopiclone is a non-benzodiazepine hypnotic sold under the trade names Imovane™ and Zimovane and is prescribed for the treatment of insomnia.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8, 1435933-78-0
Linear Formula C17H13ClN6O3D4
MDL Number MFCD16295185
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 392.83
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.