Learn More
CERILLO INC Co-Culture Duet Systems 10x (Sterilized 10 Frames, 10 Tray and 180 duets)
Catalog No. NC2389319
Encompass_Preferred
Cerillo Co-Culture System allows you to observe and study individual microbial populations in real-time. Measure the growth of one microbial population alongside other populations.
Catalog No. NC2389319 Supplier CERILLO INC Supplier No. CCU1000
Add to Cart
Add to Cart
Request a Quote
Cerillo Co-Culture System allows you to observe and study individual microbial populations in real-time. Measure the growth of one microbial population alongside other populations.