Learn More
Medchemexpress LLC HY-15656 10mg , Ceritinib CAS:1032900-25-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1565610MG
Encompass_Preferred
HY-15656 10mg Ceritinib CAS: 1032900-25-6 Ceritinib (LDK378) is a selective, orally bioavailable, and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib (LDK378) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib (LDK378) shows great antitumor potency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0933