Learn More
Selleck Chemical LLC Ceritinib
SDP
Catalog No. 5000000947
Encompass_Preferred
Ceritinib is a potent inhibitor against ALK with IC50 of 0 2 nM in cell-free assays Ceritinib (LDK378) also inhibits IGF-1R InsR STK22D and FLT3 with IC50 of 8 nM 7 nM 23 nM and 60 nM respectively Phase 3
Catalog No. 50-000-00947 Supplier Selleck Chemical LLC Supplier No. S7083200MG
Ceritinib is a potent inhibitor against ALK with IC50 of 0 2 nM in cell-free assays Ceritinib (LDK378) also inhibits IGF-1R InsR STK22D and FLT3 with IC50 of 8 nM 7 nM 23 nM and 60 nM respectively Phase 3