Learn More
Selleck Chemical LLC Ceritinib dihydrochloride-5mg
SDP
Catalog No. 502390021
Encompass_Preferred
Ceritinib (Zykadia, LDK378) dihydrochloride is a selective, orally bioavailable and ATP-competitive inhibitor of ALK with IC50 of 0.2 nM. Ceritinib dihydrochloride also inhibits InsR, IGF-1R and STK22D with IC50 of 7 nM, 8 nM and 23 nM, respectively. Ceritinib exhibits antitomor activity.
Catalog No. 50-239-0021 Supplier Selleck Chemical LLC Supplier No. S49675MG
Ceritinib (Zykadia, LDK378) dihydrochloride is a selective, orally bioavailable and ATP-competitive inhibitor of ALK with IC50 of 0.2 nM. Ceritinib dihydrochloride also inhibits InsR, IGF-1R and STK22D with IC50 of 7 nM, 8 nM and 23 nM, respectively. Ceritinib exhibits antitomor activity.