Learn More
Enzo Life Sciences Ceritinib (free base) (10mg). CAS: 1032900-25-6
SDP
Catalog No. 502013563
Encompass_Preferred
Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3563 Supplier Enzo Life Sciences Supplier No. ENZCHM2180010
Add to Cart
Add to Cart
Ceritinib is a selective inhibitor of ALK, a target found in metastatic non-small cell lung cancer. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.