Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) 2,4-pentanedionate hydrate

Catalog No. AA2111622
Click to view available options
Quantity:
100 g
500 g

Catalog No. AA2111622

Supplier: Thermo Scientific Chemicals 02111622

Only null left
Add to Cart

CAS: 206996-61-4 | C15H21CeO6 | 437.44 g/mol

Cerium(III) 2,4-pentanedionate hydrate used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206996-61-4
Molecular Formula C15H21CeO6
Molecular Weight (g/mol) 437.44
MDL Number MFCD00150164
InChI Key LCRQZRNVLFDPAZ-UHFFFAOYSA-N
Synonym cerium iii acetylacetonate hydrate, cerium iii 2,4-pentanedionate, cerium iii 2,4-pentanedionate hydrate, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy cerio oxy pent-3-en-2-one hydrate
PubChem CID 16212434
SMILES [Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 131°C to 132°C
Color Red to Yellow
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name cerium;(Z)-4-hydroxypent-3-en-2-one;hydrate
Formula Weight 437.44 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) 2,4-pentanedionate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-720-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.